missing translation for 'onlineSavingsMsg'
Learn More

4-(Chloromethyl)pyridine Hydrochloride 98.0+%, TCI America™

Supplier:  TCI America C0805250G

 View more versions of this product

Catalog No. C0805250G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

4-(Chloromethyl)pyridine Hydrochloride
169°C
C6H7Cl2N
250 g
4-chloromethyl pyridine hydrochloride, 4-picolyl chloride hydrochloride, 4-chloromethyl pyridinium chloride, 4-chloromethylpyridine hydrochloride, 4-chloromethyl-pyridine hydrochloride, ccris 1728, isonicotinyl chloride hydrochloride, pyridine, 4-chloromethyl-, hydrochloride, 4-chloromethyl pyridine hcl, 4-chloromethyl pyridine, chloride
C1=CN=CC=C1CCl.Cl
164.029
164.03
Crystalline Powder
1822-51-1
Yellow
MFCD00012826
3261
ZDHKVKPZQKYREU-UHFFFAOYSA-N
4-(chloromethyl)pyridine;hydrochloride
74570
≥98.0% (HPLC,T)

Chemical Identifiers

1822-51-1
164.029
ZDHKVKPZQKYREU-UHFFFAOYSA-N
74570
C1=CN=CC=C1CCl.Cl
C6H7Cl2N
MFCD00012826
4-chloromethyl pyridine hydrochloride, 4-picolyl chloride hydrochloride, 4-chloromethyl pyridinium chloride, 4-chloromethylpyridine hydrochloride, 4-chloromethyl-pyridine hydrochloride, ccris 1728, isonicotinyl chloride hydrochloride, pyridine, 4-chloromethyl-, hydrochloride, 4-chloromethyl pyridine hcl, 4-chloromethyl pyridine, chloride
4-(chloromethyl)pyridine;hydrochloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions