missing translation for 'onlineSavingsMsg'
Learn More

4-(Chloromethyl)pyridine Hydrochloride 98.0+%, TCI America™

Catalog No. p-7124619
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C080525G 25 g
C0805250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C080525G Supplier TCI America Supplier No. C080525G
Only null left

Chemical Identifiers

CAS 1822-51-1
Molecular Formula C6H7Cl2N
Molecular Weight (g/mol) 164.029
MDL Number MFCD00012826
InChI Key ZDHKVKPZQKYREU-UHFFFAOYSA-N
Synonym 4-chloromethyl pyridine hydrochloride, 4-picolyl chloride hydrochloride, 4-chloromethyl pyridinium chloride, 4-chloromethylpyridine hydrochloride, 4-chloromethyl-pyridine hydrochloride, ccris 1728, isonicotinyl chloride hydrochloride, pyridine, 4-chloromethyl-, hydrochloride, 4-chloromethyl pyridine hcl, 4-chloromethyl pyridine, chloride
PubChem CID 74570
IUPAC Name 4-(chloromethyl)pyridine;hydrochloride
SMILES C1=CN=CC=C1CCl.Cl

Specifications

Melting Point 169°C
Color Yellow
Quantity 25 g
UN Number 3261
Formula Weight 164.03
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(Chloromethyl)pyridine Hydrochloride