accessibility menu, dialog, popup

UserName

HClClN

CAS RN 1822-51-1

IUPAC Name: hydrogen 4-(chloromethyl)pyridine chloride
Synonyms: hydrogen 4-(chloromethyl)pyridine chloride
Molecular Weight (g/mol): 164.03
Molecular Formula: C6H7Cl2N
InChi Key: ZDHKVKPZQKYREU-UHFFFAOYSA-N
SMILES: [H+].[Cl-].ClCC1=CC=NC=C1
Molecular Weight (g/mol) 
  • (5)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (5)
  • (2)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)