missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4'-Chlorochalcone 98.0+%, TCI America™
$112.81 - $350.05
Chemical Identifiers
CAS | 956-02-5 |
---|---|
Molecular Formula | C15H11ClO |
Molecular Weight (g/mol) | 242.702 |
MDL Number | MFCD00016343 |
InChI Key | HIINIOLNGCQCSM-IZZDOVSWSA-N |
Synonym | 4'-chlorochalcone, 1-4-chlorophenyl-3-phenylprop-2-en-1-one, 2e-1-4-chlorophenyl-3-phenylprop-2-en-1-one, e-1-4-chlorophenyl-3-phenylprop-2-en-1-one, chalcone, 4'-chloro, 2-propen-1-one, 1-4-chlorophenyl-3-phenyl, ketone, p-chlorophenyl styryl, benzylidene p-chloroacetophenone, 4'-chlorchalkon, e-4'-chlorochalcone |
PubChem CID | 5377008 |
IUPAC Name | (E)-1-(4-chlorophenyl)-3-phenylprop-2-en-1-one |
SMILES | C1=CC=C(C=C1)C=CC(=O)C2=CC=C(C=C2)Cl |
Chemical Identifiers
956-02-5 | |
242.702 | |
HIINIOLNGCQCSM-IZZDOVSWSA-N | |
5377008 | |
C1=CC=C(C=C1)C=CC(=O)C2=CC=C(C=C2)Cl |
C15H11ClO | |
MFCD00016343 | |
4'-chlorochalcone, 1-4-chlorophenyl-3-phenylprop-2-en-1-one, 2e-1-4-chlorophenyl-3-phenylprop-2-en-1-one, e-1-4-chlorophenyl-3-phenylprop-2-en-1-one, chalcone, 4'-chloro, 2-propen-1-one, 1-4-chlorophenyl-3-phenyl, ketone, p-chlorophenyl styryl, benzylidene p-chloroacetophenone, 4'-chlorchalkon, e-4'-chlorochalcone | |
(E)-1-(4-chlorophenyl)-3-phenylprop-2-en-1-one |
Specifications
956-02-5 | |
Yellow | |
MFCD00016343 | |
4'-chlorochalcone, 1-4-chlorophenyl-3-phenylprop-2-en-1-one, 2e-1-4-chlorophenyl-3-phenylprop-2-en-1-one, e-1-4-chlorophenyl-3-phenylprop-2-en-1-one, chalcone, 4'-chloro, 2-propen-1-one, 1-4-chlorophenyl-3-phenyl, ketone, p-chlorophenyl styryl, benzylidene p-chloroacetophenone, 4'-chlorchalkon, e-4'-chlorochalcone | |
C1=CC=C(C=C1)C=CC(=O)C2=CC=C(C=C2)Cl | |
242.702 | |
242.70 | |
Crystalline Powder |
97°C | |
C15H11ClO | |
5 g | |
HIINIOLNGCQCSM-IZZDOVSWSA-N | |
(E)-1-(4-chlorophenyl)-3-phenylprop-2-en-1-one | |
5377008 | |
≥98.0% (GC) | |
4′-Chlorochalcone |
Safety and Handling
TSCA : No