missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4'-Chlorochalcone 98.0+%, TCI America™
Supplier: TCI America C325925G
Specifications
| 4′-Chlorochalcone | |
| 97°C | |
| C15H11ClO | |
| 25 g | |
| HIINIOLNGCQCSM-IZZDOVSWSA-N | |
| (E)-1-(4-chlorophenyl)-3-phenylprop-2-en-1-one | |
| 5377008 | |
| ≥98.0% (GC) |
| 956-02-5 | |
| Yellow | |
| MFCD00016343 | |
| 4'-chlorochalcone, 1-4-chlorophenyl-3-phenylprop-2-en-1-one, 2e-1-4-chlorophenyl-3-phenylprop-2-en-1-one, e-1-4-chlorophenyl-3-phenylprop-2-en-1-one, chalcone, 4'-chloro, 2-propen-1-one, 1-4-chlorophenyl-3-phenyl, ketone, p-chlorophenyl styryl, benzylidene p-chloroacetophenone, 4'-chlorchalkon, e-4'-chlorochalcone | |
| C1=CC=C(C=C1)C=CC(=O)C2=CC=C(C=C2)Cl | |
| 242.702 | |
| 242.70 | |
| Crystalline Powder |
Chemical Identifiers
| 956-02-5 | |
| 242.702 | |
| HIINIOLNGCQCSM-IZZDOVSWSA-N | |
| 5377008 | |
| C1=CC=C(C=C1)C=CC(=O)C2=CC=C(C=C2)Cl |
| C15H11ClO | |
| MFCD00016343 | |
| 4'-chlorochalcone, 1-4-chlorophenyl-3-phenylprop-2-en-1-one, 2e-1-4-chlorophenyl-3-phenylprop-2-en-1-one, e-1-4-chlorophenyl-3-phenylprop-2-en-1-one, chalcone, 4'-chloro, 2-propen-1-one, 1-4-chlorophenyl-3-phenyl, ketone, p-chlorophenyl styryl, benzylidene p-chloroacetophenone, 4'-chlorchalkon, e-4'-chlorochalcone | |
| (E)-1-(4-chlorophenyl)-3-phenylprop-2-en-1-one |
Safety and Handling
TSCA : No