missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chlorobutyl Acetate 97.0+%, TCI America™
Supplier: TCI America A1098250G
Specifications
| 4-Chlorobutyl Acetate | |
| Colorless | |
| C6H11ClO2 | |
| 250 g | |
| PYLDCZJUHYVOAF-UHFFFAOYSA-N | |
| 4-chlorobutyl acetate | |
| 23399 | |
| ≥97.0% (GC) |
| 6962-92-1 | |
| 87°C | |
| MFCD00001013 | |
| 1-acetoxy-4-chlorobutane, 4-chloro-n-butyl acetate, 1-chloro-4-acetoxybutane, 1-butanol, 4-chloro-, acetate, 4-chlorobutylacetate, 4-chloro-1-butanol acetate, 4-chlorbut-1-ylacetat, delta-chlorobutyl acetate, 1-butanol, 4-chloro-, 1-acetate, 4-chlorbut-1-ylacetat german | |
| CC(=O)OCCCCCl | |
| 150.602 | |
| 150.60 | |
| Liquid |
Chemical Identifiers
| 6962-92-1 | |
| 150.602 | |
| PYLDCZJUHYVOAF-UHFFFAOYSA-N | |
| 23399 | |
| CC(=O)OCCCCCl |
| C6H11ClO2 | |
| MFCD00001013 | |
| 1-acetoxy-4-chlorobutane, 4-chloro-n-butyl acetate, 1-chloro-4-acetoxybutane, 1-butanol, 4-chloro-, acetate, 4-chlorobutylacetate, 4-chloro-1-butanol acetate, 4-chlorbut-1-ylacetat, delta-chlorobutyl acetate, 1-butanol, 4-chloro-, 1-acetate, 4-chlorbut-1-ylacetat german | |
| 4-chlorobutyl acetate |
Safety and Handling
EINECSNumber : (2)-0737
RTECSNumber : EL1328000
TSCA : Yes