accessibility menu, dialog, popup

UserName

H3COOCl

CAS RN 6962-92-1

IUPAC Name: 4-chlorobutyl acetate
Synonyms: 1-butanol, 4-chloro-, acetate
Molecular Weight (g/mol): 150.6
Molecular Formula: C6H11ClO2
InChi Key: PYLDCZJUHYVOAF-UHFFFAOYSA-N
SMILES: CC(=O)OCCCCCl
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)