missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobutyl Acetate 97.0+%, TCI America™

Catalog No. p-7117243
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A109825G 25 g
A1098250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A109825G Supplier TCI America Supplier No. A109825G
Only null left

Chemical Identifiers

CAS 6962-92-1
Molecular Formula C6H11ClO2
Molecular Weight (g/mol) 150.602
MDL Number MFCD00001013
InChI Key PYLDCZJUHYVOAF-UHFFFAOYSA-N
Synonym 1-acetoxy-4-chlorobutane, 4-chloro-n-butyl acetate, 1-chloro-4-acetoxybutane, 1-butanol, 4-chloro-, acetate, 4-chlorobutylacetate, 4-chloro-1-butanol acetate, 4-chlorbut-1-ylacetat, delta-chlorobutyl acetate, 1-butanol, 4-chloro-, 1-acetate, 4-chlorbut-1-ylacetat german
PubChem CID 23399
IUPAC Name 4-chlorobutyl acetate
SMILES CC(=O)OCCCCCl

Specifications

Color Colorless
Boiling Point 87°C
Quantity 25 g
Formula Weight 150.60
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Chlorobutyl Acetate
EINECSNumber (2)-0737
RTECSNumber EL1328000
TSCA Yes