missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chlorobenzyl Cyanide 98.0+%, TCI America™
$32.72 - $186.74
Chemical Identifiers
| CAS | 140-53-4 |
|---|---|
| Molecular Formula | C8H6ClN |
| Molecular Weight (g/mol) | 151.593 |
| MDL Number | MFCD00001918 |
| InChI Key | IVYMIRMKXZAHRV-UHFFFAOYSA-N |
| Synonym | 4-chlorobenzyl cyanide, 4-chlorophenyl acetonitrile, 4-chlorophenylacetonitrile, 2-4-chlorophenyl acetonitrile, p-chlorobenzyl cyanide, 4-chlorobenzeneacetonitrile, benzeneacetonitrile, 4-chloro, p-chlorophenyl acetonitrile, p-chlorophenylacetonitrile, 4-chlorobenzylcyanide |
| PubChem CID | 241582 |
| ChEBI | CHEBI:17346 |
| IUPAC Name | 2-(4-chlorophenyl)acetonitrile |
| SMILES | C1=CC(=CC=C1CC#N)Cl |
Chemical Identifiers
| 140-53-4 | |
| 151.593 | |
| IVYMIRMKXZAHRV-UHFFFAOYSA-N | |
| 241582 | |
| 2-(4-chlorophenyl)acetonitrile |
| C8H6ClN | |
| MFCD00001918 | |
| 4-chlorobenzyl cyanide, 4-chlorophenyl acetonitrile, 4-chlorophenylacetonitrile, 2-4-chlorophenyl acetonitrile, p-chlorobenzyl cyanide, 4-chlorobenzeneacetonitrile, benzeneacetonitrile, 4-chloro, p-chlorophenyl acetonitrile, p-chlorophenylacetonitrile, 4-chlorobenzylcyanide | |
| CHEBI:17346 | |
| C1=CC(=CC=C1CC#N)Cl |
Specifications
| 140-53-4 | |
| 267°C | |
| MFCD00001918 | |
| 3439 | |
| IVYMIRMKXZAHRV-UHFFFAOYSA-N | |
| 2-(4-chlorophenyl)acetonitrile | |
| 241582 | |
| 151.59 | |
| Crystalline Lumps |
| White-Yellow | |
| C8H6ClN | |
| 25 g | |
| 4-chlorobenzyl cyanide, 4-chlorophenyl acetonitrile, 4-chlorophenylacetonitrile, 2-4-chlorophenyl acetonitrile, p-chlorobenzyl cyanide, 4-chlorobenzeneacetonitrile, benzeneacetonitrile, 4-chloro, p-chlorophenyl acetonitrile, p-chlorophenylacetonitrile, 4-chlorobenzylcyanide | |
| C1=CC(=CC=C1CC#N)Cl | |
| 151.593 | |
| CHEBI:17346 | |
| ≥98.0% (GC) | |
| 4-Chlorobenzyl Cyanide |
Safety and Handling
EINECSNumber : (3)-1784
RTECSNumber : AL8250000
TSCA : Yes