missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobenzyl cyanide, 98+%

Catalog No. AC108810050
Change view
Click to view available options
Quantity:
5 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC108810050 5 g Glass bottle
AC108811000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC108810050 Supplier Thermo Scientific Chemicals Supplier No. 108810050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 140-53-4 | C8H6ClN | 151.6 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 140-53-4
Molecular Formula C8H6ClN
Molecular Weight (g/mol) 151.6
MDL Number MFCD00001918
InChI Key IVYMIRMKXZAHRV-UHFFFAOYSA-N
Synonym 4-chlorobenzyl cyanide, 4-chlorophenyl acetonitrile, 4-chlorophenylacetonitrile, 2-4-chlorophenyl acetonitrile, p-chlorobenzyl cyanide, 4-chlorobenzeneacetonitrile, benzeneacetonitrile, 4-chloro, p-chlorophenyl acetonitrile, p-chlorophenylacetonitrile, 4-chlorobenzylcyanide
PubChem CID 241582
ChEBI CHEBI:17346
IUPAC Name 2-(4-chlorophenyl)acetonitrile
SMILES C1=CC(=CC=C1CC#N)Cl

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 29°C to 30°C
Color Colorless to Yellow
Boiling Point 265°C to 267°C
Flash Point 133°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Refractive Index 1.5415 to 1.5435
Linear Formula ClC6H4CH2CN
Quantity 5 g
Beilstein 09, 448
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in acetone and alcohol
Formula Weight 151.6
Percent Purity 98+%
Physical Form Liquid After Melting
Chemical Name or Material 4-Chlorobenzyl cyanide, 98+%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause respiratory irritation.
Toxic in contact with skin.
Toxic if inhaled.
Fatal if swallowed.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES:
EINECSNumber 205-418-9
RTECSNumber AL8250000
TSCA TSCA

RUO – Research Use Only