missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobenzotrifluoride 98.0+%, TCI America™

Catalog No. c030525g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
500 g
Catalog No. Quantity
C030525G 25 g
C0305500G 500 g
2 options

Catalog No. C030525G

Supplier: TCI America C030525G

Only null left

Chemical Identifiers

CAS 98-56-6
Molecular Formula C7H4ClF3
Molecular Weight (g/mol) 180.554
MDL Number MFCD00000627
InChI Key QULYNCCPRWKEMF-UHFFFAOYSA-N
Synonym 4-chlorobenzotrifluoride, p-chlorobenzotrifluoride, 1-chloro-4-trifluoromethyl benzene, benzene, 1-chloro-4-trifluoromethyl, 4-chloro-alpha,alpha,alpha-trifluorotoluene, para-chlorobenzotrifluoride, p-chlorophenyl trifluoromethane, p-chloro-a,a,a-trifluorotoluene, pcbtf, p-chlorotrifluoromethylbenzene
PubChem CID 7394
IUPAC Name 1-chloro-4-(trifluoromethyl)benzene
SMILES C1=CC(=CC=C1C(F)(F)F)Cl

Specifications

Color Colorless
Boiling Point 140°C
Quantity 25 g
UN Number 2234
Formula Weight 180.55
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Chlorobenzotrifluoride
EINECSNumber (3)-0053
RTECSNumber XS9145000
TSCA Yes
Recommended Storage Refrigerator