missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobenzotrifluoride, 98%

CAS: 98-56-6 | C7H4ClF3 | 180.56 g/mol

$44.93 - $147.84

Chemical Identifiers

CAS 98-56-6
Molecular Formula C7H4ClF3
Molecular Weight (g/mol) 180.56
MDL Number MFCD00000627
InChI Key QULYNCCPRWKEMF-UHFFFAOYSA-N
Synonym 4-chlorobenzotrifluoride, p-chlorobenzotrifluoride, 1-chloro-4-trifluoromethyl benzene, benzene, 1-chloro-4-trifluoromethyl, 4-chloro-alpha,alpha,alpha-trifluorotoluene, para-chlorobenzotrifluoride, p-chlorophenyl trifluoromethane, p-chloro-a,a,a-trifluorotoluene, pcbtf, p-chlorotrifluoromethylbenzene
PubChem CID 7394
IUPAC Name 1-chloro-4-(trifluoromethyl)benzene
SMILES C1=CC(=CC=C1C(F)(F)F)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC108730250
SDS View Documents
Thermo Scientific Chemicals
108730250
25 mL Glass bottle N/A
Only null left
 
AC108732500
SDS View Documents
Thermo Scientific Chemicals
108732500
250 mL Glass bottle N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

98-56-6
180.56
QULYNCCPRWKEMF-UHFFFAOYSA-N
7394
C1=CC(=CC=C1C(F)(F)F)Cl
C7H4ClF3
MFCD00000627
4-chlorobenzotrifluoride, p-chlorobenzotrifluoride, 1-chloro-4-trifluoromethyl benzene, benzene, 1-chloro-4-trifluoromethyl, 4-chloro-alpha,alpha,alpha-trifluorotoluene, para-chlorobenzotrifluoride, p-chlorophenyl trifluoromethane, p-chloro-a,a,a-trifluorotoluene, pcbtf, p-chlorotrifluoromethylbenzene
1-chloro-4-(trifluoromethyl)benzene

Specifications

98-56-6
100.0
Colorless
136.0°C to 138.0°C
Authentic
Glass bottle
1.4453 to 1.4473
MFCD00000627
1.35
4-chlorobenzotrifluoride, p-chlorobenzotrifluoride, 1-chloro-4-trifluoromethyl benzene, benzene, 1-chloro-4-trifluoromethyl, 4-chloro-alpha,alpha,alpha-trifluorotoluene, para-chlorobenzotrifluoride, p-chlorophenyl trifluoromethane, p-chloro-a,a,a-trifluorotoluene, pcbtf, p-chlorotrifluoromethylbenzene
C1=CC(=CC=C1C(F)(F)F)Cl
180.56
0.67 mPa.s (38°C)
98%
4-Chlorobenzotrifluoride, 98%
97.5
-36.0°C
1.3500g/mL
47°C
98%
C7H4ClF3
ClC6H4CF3
25 mL
15, 2127
QULYNCCPRWKEMF-UHFFFAOYSA-N
1-chloro-4-(trifluoromethyl)benzene
7394
180.56
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Toxic to aquatic life with long lasting effects.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wash face,hands and any exposed skin thorou

GHS Signal Word: Danger

EINECSNumber : 202-681-1

RTECSNumber : XS9145000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only