missing translation for 'onlineSavingsMsg'
Learn More

4'-Chlorobenzophenone-2-carboxylic Acid 98.0+%, TCI America™

Catalog No. C107425G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C107425G 25 g
C1074500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C107425G Supplier TCI America Supplier No. C107425G

Chemical Identifiers

CAS 85-56-3
Molecular Formula C14H9ClO3
Molecular Weight (g/mol) 260.673
MDL Number MFCD00002474
InChI Key YWECCEXWKFHHQJ-UHFFFAOYSA-N
Synonym 2-4-chlorobenzoyl benzoic acid, benzoic acid, 2-4-chlorobenzoyl, o-p-chlorobenzoyl benzoic acid, 2-p-chlorobenzoyl benzoic acid, o-4-chlorobenzoyl benzoic acid, 2-4'-chlorobenzoyl benzoic acid, unii-3u7w4yoy3r, 3u7w4yoy3r, chembl81936, 2-4-chloro-benzoyl-benzoic acid
PubChem CID 66564
IUPAC Name 2-(4-chlorobenzoyl)benzoic acid
SMILES C1=CC=C(C(=C1)C(=O)C2=CC=C(C=C2)Cl)C(=O)O

Specifications

Melting Point 151°C
Color White
Quantity 25 g
Formula Weight 260.67
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Chlorobenzophenone-2-carboxylic Acid
EINECSNumber (4)-0137
TSCA Yes