missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chlorobenzoic Acid 99.0+%, TCI America™
$41.02 - $152.86
Chemical Identifiers
| CAS | 74-11-3 |
|---|---|
| Molecular Formula | C7H5ClO2 |
| Molecular Weight (g/mol) | 156.565 |
| MDL Number | MFCD00002531 |
| InChI Key | XRHGYUZYPHTUJZ-UHFFFAOYSA-N |
| Synonym | p-chlorobenzoic acid, benzoic acid, 4-chloro, p-chlorbenzoic acid, chlorodracylic acid, benzoic acid, p-chloro, para-chlorobenzoic acid, 4-chloro-benzoic acid, p-carboxychlorobenzene, acido p-clorobenzoico, acido p-clorobenzoico italian |
| PubChem CID | 6318 |
| ChEBI | CHEBI:30747 |
| IUPAC Name | 4-chlorobenzoic acid |
| SMILES | C1=CC(=CC=C1C(=O)O)Cl |
Chemical Identifiers
| 74-11-3 | |
| 156.565 | |
| XRHGYUZYPHTUJZ-UHFFFAOYSA-N | |
| 6318 | |
| 4-chlorobenzoic acid |
| C7H5ClO2 | |
| MFCD00002531 | |
| p-chlorobenzoic acid, benzoic acid, 4-chloro, p-chlorbenzoic acid, chlorodracylic acid, benzoic acid, p-chloro, para-chlorobenzoic acid, 4-chloro-benzoic acid, p-carboxychlorobenzene, acido p-clorobenzoico, acido p-clorobenzoico italian | |
| CHEBI:30747 | |
| C1=CC(=CC=C1C(=O)O)Cl |
Specifications
| 74-11-3 | |
| White | |
| MFCD00002531 | |
| p-chlorobenzoic acid, benzoic acid, 4-chloro, p-chlorbenzoic acid, chlorodracylic acid, benzoic acid, p-chloro, para-chlorobenzoic acid, 4-chloro-benzoic acid, p-carboxychlorobenzene, acido p-clorobenzoico, acido p-clorobenzoico italian | |
| C1=CC(=CC=C1C(=O)O)Cl | |
| 156.565 | |
| CHEBI:30747 | |
| ≥99.0% (GC,T) | |
| 4-Chlorobenzoic Acid |
| 242°C | |
| C7H5ClO2 | |
| 25 g | |
| XRHGYUZYPHTUJZ-UHFFFAOYSA-N | |
| 4-chlorobenzoic acid | |
| 6318 | |
| 156.57 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (3)-0062&(3)-1424
RTECSNumber : DG4976010
TSCA : Yes