missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobenzoic Acid 99.0+%, TCI America™

Supplier:  TCI America C0134500G

 View more versions of this product

Catalog No. C0134500G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

4-Chlorobenzoic Acid
242°C
C7H5ClO2
500 g
XRHGYUZYPHTUJZ-UHFFFAOYSA-N
4-chlorobenzoic acid
6318
156.57
Crystalline Powder
74-11-3
White
MFCD00002531
p-chlorobenzoic acid, benzoic acid, 4-chloro, p-chlorbenzoic acid, chlorodracylic acid, benzoic acid, p-chloro, para-chlorobenzoic acid, 4-chloro-benzoic acid, p-carboxychlorobenzene, acido p-clorobenzoico, acido p-clorobenzoico italian
C1=CC(=CC=C1C(=O)O)Cl
156.565
CHEBI:30747
≥99.0% (GC,T)

Chemical Identifiers

74-11-3
156.565
XRHGYUZYPHTUJZ-UHFFFAOYSA-N
6318
4-chlorobenzoic acid
C7H5ClO2
MFCD00002531
p-chlorobenzoic acid, benzoic acid, 4-chloro, p-chlorbenzoic acid, chlorodracylic acid, benzoic acid, p-chloro, para-chlorobenzoic acid, 4-chloro-benzoic acid, p-carboxychlorobenzene, acido p-clorobenzoico, acido p-clorobenzoico italian
CHEBI:30747
C1=CC(=CC=C1C(=O)O)Cl
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0062&(3)-1424

RTECSNumber : DG4976010

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions