Learn More
4-Chlorobenzoic acid, 98+%
CAS: 74-11-3 | C7H5ClO2 | 156.565 g/mol
Supplier: Thermo Scientific Chemicals A151350B
Description
4-Chlorobenzoic acid is used to determine the diversity of 2-, 3- and 4-chlorobenzoate degraders in two pristine soils with similar physical and chemical characteristics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Chlorobenzoic acid is used to determine the diversity of 2-, 3- and 4-chlorobenzoate degraders in two pristine soils with similar physical and chemical characteristics.
Solubility
Soluble in water(0.08g/L).
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
| 4-Chlorobenzoic acid | |
| 239°C to 243°C | |
| 275°C | |
| Odorless | |
| MFCD00002531 | |
| 907196 | |
| p-chlorobenzoic acid, benzoic acid, 4-chloro, p-chlorbenzoic acid, chlorodracylic acid, benzoic acid, p-chloro, para-chlorobenzoic acid, 4-chloro-benzoic acid, p-carboxychlorobenzene, acido p-clorobenzoico, acido p-clorobenzoico italian | |
| XRHGYUZYPHTUJZ-UHFFFAOYSA-N | |
| 4-chlorobenzoic acid | |
| 6318 | |
| 156.57 |
| 74-11-3 | |
| 1.54 | |
| 238°C (460°F) | |
| C7H5ClO2 | |
| 1000 g | |
| 14,2125 | |
| Soluble in water(0.08g/L). | |
| C1=CC(=CC=C1C(=O)O)Cl | |
| 156.565 | |
| CHEBI:30747 | |
| ≥98% |
Chemical Identifiers
| 74-11-3 | |
| 156.565 | |
| XRHGYUZYPHTUJZ-UHFFFAOYSA-N | |
| 6318 | |
| 4-chlorobenzoic acid |
| C7H5ClO2 | |
| MFCD00002531 | |
| p-chlorobenzoic acid, benzoic acid, 4-chloro, p-chlorbenzoic acid, chlorodracylic acid, benzoic acid, p-chloro, para-chlorobenzoic acid, 4-chloro-benzoic acid, p-carboxychlorobenzene, acido p-clorobenzoico, acido p-clorobenzoico italian | |
| CHEBI:30747 | |
| C1=CC(=CC=C1C(=O)O)Cl |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 200-805-9
RTECSNumber : DG4976010
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only