missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobenzoic acid, 98+%

Catalog No. p-7022385
Change view
Click to view available options
Quantity:
250 g
1000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1513530 250 g
AAA151350B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1513530 Supplier Thermo Scientific Chemicals Supplier No. A1513530
Only null left

CAS: 74-11-3 | C7H5ClO2 | 156.565 g/mol

4-Chlorobenzoic acid is used to determine the diversity of 2-, 3- and 4-chlorobenzoate degraders in two pristine soils with similar physical and chemical characteristics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chlorobenzoic acid is used to determine the diversity of 2-, 3- and 4-chlorobenzoate degraders in two pristine soils with similar physical and chemical characteristics.

Solubility
Soluble in water(0.08g/L).

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 74-11-3
Molecular Formula C7H5ClO2
Molecular Weight (g/mol) 156.565
MDL Number MFCD00002531
InChI Key XRHGYUZYPHTUJZ-UHFFFAOYSA-N
Synonym p-chlorobenzoic acid, benzoic acid, 4-chloro, p-chlorbenzoic acid, chlorodracylic acid, benzoic acid, p-chloro, para-chlorobenzoic acid, 4-chloro-benzoic acid, p-carboxychlorobenzene, acido p-clorobenzoico, acido p-clorobenzoico italian
PubChem CID 6318
ChEBI CHEBI:30747
IUPAC Name 4-chlorobenzoic acid
SMILES C1=CC(=CC=C1C(=O)O)Cl

Specifications

Melting Point 239°C to 243°C
Density 1.54
Boiling Point 275°C
Flash Point 238°C (460°F)
Odor Odorless
Quantity 250 g
Beilstein 907196
Merck Index 14,2125
Solubility Information Soluble in water(0.08g/L).
Formula Weight 156.57
Percent Purity ≥98%
Chemical Name or Material 4-Chlorobenzoic acid
Show More Show Less
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 200-805-9
RTECSNumber DG4976010
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only