missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobenzhydrol, 98%

Catalog No. p-7027153
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2369106 5 g
AAB2369114 25 g
AAB2369122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2369106 Supplier Thermo Scientific Chemicals Supplier No. B2369106

CAS: 119-56-2 | C13H11ClO | 218.68 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 119-56-2
Molecular Formula C13H11ClO
Molecular Weight (g/mol) 218.68
MDL Number MFCD00004491
InChI Key AJYOOHCNOXWTKJ-UHFFFAOYSA-N
Synonym 4-chlorobenzhydrol, 4-chlorophenyl phenyl methanol, p-chlorobenzhydrol, chlorobenzhydrol, benzhydrol, p-chloro, 4-chlorodiphenylmethanol, benzhydrol, 4-chloro, 4-chlorophenyl phenylmethanol, 4-chlorobenzhydryl alcohol, benzenemethanol, 4-chloro-.alpha.-phenyl
PubChem CID 8401
ChEBI CHEBI:35091
IUPAC Name (4-chlorophenyl)-phenylmethanol
SMILES C1=CC=C(C=C1)C(C2=CC=C(C=C2)Cl)O

Specifications

Melting Point 58°C to 62°C
Boiling Point 150°C to 151°C (2.6 mmHg)
Quantity 5 g
Beilstein 2050235
Formula Weight 218.68
Percent Purity 98%
Chemical Name or Material 4-Chlorobenzhydrol
EINECSNumber 204-333-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only