accessibility menu, dialog, popup

UserName

HOCl

CAS RN 119-56-2

IUPAC Name: (4-chlorophenyl)(phenyl)methanol
Synonyms: chlorobenzhydrol C13H11ClO
Molecular Weight (g/mol): 218.68
Molecular Formula: C13H11ClO
InChi Key: AJYOOHCNOXWTKJ-UHFFFAOYNA-N
SMILES: OC(C1=CC=CC=C1)C1=CC=C(Cl)C=C1
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)