missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobenzenesulfonamide 98.0+%, TCI America™

Catalog No. C1099500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C1099500G 500 g
C109925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C1099500G Supplier TCI America Supplier No. C1099500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 98-64-6
Molecular Formula C6H6ClNO2S
Molecular Weight (g/mol) 191.63
MDL Number MFCD00007936
InChI Key HHHDJHHNEURCNV-UHFFFAOYSA-N
Synonym p-chlorobenzenesulfonamide, benzenesulfonamide, 4-chloro, p-chlorobenzenesulfamide, 4-chlorophenylsulfonamide, p-chlorophenylsulfonamide, 4-chlorobenzenesulphonamide, benzenesulfonamide, p-chloro, usaf ma-3, 4-chlorobenzene-1-sulfonamide, 4-chloro-benzenesulfonamide
PubChem CID 66824
IUPAC Name 4-chlorobenzene-1-sulfonamide
SMILES NS(=O)(=O)C1=CC=C(Cl)C=C1

Specifications

Melting Point 144°C
Color White
Quantity 500 g
Formula Weight 191.63
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chlorobenzenesulfonamide
EINECSNumber (3)-1954
RTECSNumber DB1400000
TSCA Yes