accessibility menu, dialog, popup

UserName

NH2SOOCl

CAS RN 98-64-6

IUPAC Name: 4-chlorobenzene-1-sulfonamide
Synonyms: 4-chlorobenzenesulfonamide
Molecular Weight (g/mol): 191.63
Molecular Formula: C6H6ClNO2S
InChi Key: HHHDJHHNEURCNV-UHFFFAOYSA-N
SMILES: NS(=O)(=O)C1=CC=C(Cl)C=C1
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)