missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobenzamide 98.0+%, TCI America™

Catalog No. p-7124500
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C056325G 25 g
C0563250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C056325G Supplier TCI America Supplier No. C056325G
Only null left

Chemical Identifiers

CAS 619-56-7
Molecular Formula C7H6ClNO
Molecular Weight (g/mol) 155.58
MDL Number MFCD00007993
InChI Key BLNVISNJTIRAHF-UHFFFAOYSA-N
Synonym p-chlorobenzamide, benzamide, 4-chloro, 4-chloro-benzamide, p-chlorobenzoic acid amide, benzamide, p-chloro, unii-260cv6k0mr, benzamide, p-chloro-6ci,7ci,8ci, 4-chloranylbenzamide, pubchem3619, 4-chlorobenzenecarboxamide
PubChem CID 12084
IUPAC Name 4-chlorobenzamide
SMILES NC(=O)C1=CC=C(Cl)C=C1

Specifications

Melting Point 179°C
Color White
Quantity 25 g
Formula Weight 155.58
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chlorobenzamide
EINECSNumber (3)-1430
TSCA No