missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chlorobenzaldehyde 97.0+%, TCI America™
Supplier: TCI America C0125100G
Specifications
| 4-Chlorobenzaldehyde | |
| 48°C | |
| 214°C | |
| MFCD00003379 | |
| p-chlorobenzaldehyde, benzaldehyde, 4-chloro, benzaldehyde, p-chloro, p-chlorobenzenecarboxaldehyde, 4-chloro-benzaldehyde, 4-chlorobenzenaldehyde, 4-chloro benzaldehyde, ccris 857, para-chlorobenzaldehyde, unii-e67727up9z | |
| C1=CC(=CC=C1C=O)Cl | |
| 140.566 | |
| CHEBI:28105 | |
| ≥97.0% (GC) |
| 104-88-1 | |
| White-Yellow | |
| C7H5ClO | |
| 100 g | |
| AVPYQKSLYISFPO-UHFFFAOYSA-N | |
| 4-chlorobenzaldehyde | |
| 7726 | |
| 140.57 | |
| Crystalline Powder |
Chemical Identifiers
| 104-88-1 | |
| 140.566 | |
| AVPYQKSLYISFPO-UHFFFAOYSA-N | |
| 7726 | |
| 4-chlorobenzaldehyde |
| C7H5ClO | |
| MFCD00003379 | |
| p-chlorobenzaldehyde, benzaldehyde, 4-chloro, benzaldehyde, p-chloro, p-chlorobenzenecarboxaldehyde, 4-chloro-benzaldehyde, 4-chlorobenzenaldehyde, 4-chloro benzaldehyde, ccris 857, para-chlorobenzaldehyde, unii-e67727up9z | |
| CHEBI:28105 | |
| C1=CC(=CC=C1C=O)Cl |
Safety and Handling
EINECSNumber : (3)-1162
RTECSNumber : CU5076000
TSCA : Yes