missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobenzaldehyde 97.0+%, TCI America™

Catalog No. p-7124228
Change view
Click to view available options
Quantity:
100 g
25 g
500 g
Catalog No. Quantity
C012525G 25 g
C0125100G 100 g
C0125500G 500 g
3 options

Catalog No. C012525G

Supplier: TCI America C012525G

Only null left

Chemical Identifiers

CAS 104-88-1
Molecular Formula C7H5ClO
Molecular Weight (g/mol) 140.566
MDL Number MFCD00003379
InChI Key AVPYQKSLYISFPO-UHFFFAOYSA-N
Synonym p-chlorobenzaldehyde, benzaldehyde, 4-chloro, benzaldehyde, p-chloro, p-chlorobenzenecarboxaldehyde, 4-chloro-benzaldehyde, 4-chlorobenzenaldehyde, 4-chloro benzaldehyde, ccris 857, para-chlorobenzaldehyde, unii-e67727up9z
PubChem CID 7726
ChEBI CHEBI:28105
IUPAC Name 4-chlorobenzaldehyde
SMILES C1=CC(=CC=C1C=O)Cl

Specifications

Melting Point 48°C
Color White-Yellow
Boiling Point 214°C
Quantity 25 g
Formula Weight 140.57
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chlorobenzaldehyde
EINECSNumber (3)-1162
RTECSNumber CU5076000
TSCA Yes