missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-o-cresol 90.0+%, TCI America™

Catalog No. C015225G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C015225G 25 g
C0152500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C015225G Supplier TCI America Supplier No. C015225G

Chemical Identifiers

CAS 1570-64-5
Molecular Formula C7H7ClO
Molecular Weight (g/mol) 142.582
MDL Number MFCD00002321
InChI Key RHPUJHQBPORFGV-UHFFFAOYSA-N
Synonym 4-chloro-o-cresol, 2-methyl-4-chlorophenol, 4-chloro-2-cresol, p-chloro-o-cresol, phenol, 4-chloro-2-methyl, o-cresol, 4-chloro, 4-chloro-2-methyl phenol, 4-chloro-2-methyl-phenol, 5-chloro-2-hydroxytoluene, unii-297v63w9ri
PubChem CID 14855
ChEBI CHEBI:1800
IUPAC Name 4-chloro-2-methylphenol
SMILES CC1=C(C=CC(=C1)Cl)O

Specifications

Quantity 25 g
UN Number 3437
Formula Weight 142.58
Percent Purity ≥90.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 4-Chloro-o-cresol
EINECSNumber (3)-0900
RTECSNumber GO7120000
TSCA Yes