accessibility menu, dialog, popup

UserName

CH3ClHO

CAS RN 1570-64-5

IUPAC Name: 4-chloro-2-methylphenol
Synonyms: 4-chloro-2-methylphenol
Molecular Weight (g/mol): 142.58
Molecular Formula: C7H7ClO
InChi Key: RHPUJHQBPORFGV-UHFFFAOYSA-N
SMILES: CC1=CC(Cl)=CC=C1O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)