missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-N-methylbenzylamine 97.0+%, TCI America™

Catalog No. p-7125701
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C28851G 1 g
C28855G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C28851G Supplier TCI America Supplier No. C28851G
Only null left

Chemical Identifiers

CAS 104-11-0
Molecular Formula C8H10ClN
Molecular Weight (g/mol) 155.625
MDL Number MFCD00018749
InChI Key LMBUJNXYGGNSAH-UHFFFAOYSA-N
Synonym 4-chloro-n-methylbenzylamine, n-methyl-4-chlorobenzylamine, 1-4-chlorophenyl-n-methylmethanamine, n-4-chlorobenzyl-n-methylamine, 4-chloro-benzyl-methyl-amine, 4-chlorophenyl methyl methyl amine, 4-chlorobenzyl methylamine, benzenemethanamine, 4-chloro-n-methyl, 4-chlorobenzylmethylamine, p-chloro-n-methylbenzylamine
PubChem CID 66905
IUPAC Name 1-(4-chlorophenyl)-N-methylmethanamine
SMILES CNCC1=CC=C(C=C1)Cl

Specifications

Color Yellow
Boiling Point 119°C
Quantity 1 g
UN Number 2735
Formula Weight 155.63
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-Chloro-N-methylbenzylamine