accessibility menu, dialog, popup

UserName

CH3HNCl

CAS RN 104-11-0

IUPAC Name: [(4-chlorophenyl)methyl](methyl)amine
Synonyms: [(4-chlorophenyl)methyl](methyl)amine
Molecular Weight (g/mol): 155.63
Molecular Formula: C8H10ClN
InChi Key: LMBUJNXYGGNSAH-UHFFFAOYSA-N
SMILES: CNCC1=CC=C(Cl)C=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results