missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chloro-N-methylbenzylamine 97.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 119°C |
| Quantity | 5 g |
| UN Number | 2735 |
| Formula Weight | 155.63 |
| Percent Purity | ≥97.0% (GC,T) |
| Physical Form | Liquid |
| Chemical Name or Material | 4-Chloro-N-methylbenzylamine |
Safety and Handling
| TSCA | Yes |