missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chloro-DL-phenylalanine 98.0+%, TCI America™
Chemical Identifiers
| CAS | 7424-00-2 |
|---|---|
| Molecular Formula | C9H10ClNO2 |
| Molecular Weight (g/mol) | 199.63 |
| MDL Number | MFCD00002601 |
| InChI Key | NIGWMJHCCYYCSF-UHFFFAOYNA-N |
| Synonym | fenclonine, 4-chloro-dl-phenylalanine, p-chlorophenylalanine, 2-amino-3-4-chlorophenyl propanoic acid, 4-chlorophenylalanine, dl-4-chlorophenylalanine, dl-p-chlorophenylalanine, fenchlonine, fenclonin, phenylalanine, 4-chloro |
| PubChem CID | 4652 |
| IUPAC Name | 2-amino-3-(4-chlorophenyl)propanoic acid |
| SMILES | NC(CC1=CC=C(Cl)C=C1)C(O)=O |
Chemical Identifiers
| C9H10ClNO2 | |
| MFCD00002601 | |
| fenclonine, 4-chloro-dl-phenylalanine, p-chlorophenylalanine, 2-amino-3-4-chlorophenyl propanoic acid, 4-chlorophenylalanine, dl-4-chlorophenylalanine, dl-p-chlorophenylalanine, fenchlonine, fenclonin, phenylalanine, 4-chloro | |
| 2-amino-3-(4-chlorophenyl)propanoic acid |
Specifications
| 7424-00-2 | |
| C9H10ClNO2 | |
| fenclonine, 4-chloro-dl-phenylalanine, p-chlorophenylalanine, 2-amino-3-4-chlorophenyl propanoic acid, 4-chlorophenylalanine, dl-4-chlorophenylalanine, dl-p-chlorophenylalanine, fenchlonine, fenclonin, phenylalanine, 4-chloro | |
| NC(CC1=CC=C(Cl)C=C1)C(O)=O | |
| 199.63 | |
| 199.63 | |
| Crystalline Powder |
| White-Yellow | |
| MFCD00002601 | |
| NIGWMJHCCYYCSF-UHFFFAOYNA-N | |
| 2-amino-3-(4-chlorophenyl)propanoic acid | |
| 4652 | |
| ≥98.0% (T) | |
| 4-Chloro-DL-phenylalanine |
Safety and Handling
RTECSNumber : AY4390010
TSCA : Yes