accessibility menu, dialog, popup

UserName

ClNH2OOH

CAS RN 7424-00-2

IUPAC Name: 2-amino-3-(4-chlorophenyl)propanoic acid
Synonyms: fenclonine|C-pal|hydrochloride, fenclonine|DL-pcpa|fenclonina|p-clorophenylalanine|fenclonin|fenchlonine|fencloninum|fenclonine (L)-
Molecular Weight (g/mol): 199.63
Molecular Formula: C9H10ClNO2
InChi Key: NIGWMJHCCYYCSF-UHFFFAOYNA-N
SMILES: NC(CC1=CC=C(Cl)C=C1)C(O)=O
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)