missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chloro-DL-phenylalanine 98.0+%, TCI America™
Specifications
Specifications
| Color | White-Yellow |
| Quantity | 25 g |
| Formula Weight | 199.63 |
| Percent Purity | ≥98.0% (T) |
| Grade | Research |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4-Chloro-DL-phenylalanine |
Safety and Handling
| RTECSNumber | AY4390010 |
| TSCA | Yes |