missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-alpha,alpha-dimethylphenylacetic acid, 98%

CAS: 6258-30-6 | C10H11ClO2 | 198.646 g/mol

$33.90 - $113.15

Chemical Identifiers

CAS 6258-30-6
Molecular Formula C10H11ClO2
Molecular Weight (g/mol) 198.646
MDL Number MFCD00044814
InChI Key SSFDAZXGUKDEAH-UHFFFAOYSA-N
Synonym 2-4-chlorophenyl-2-methylpropanoic acid, 2-4-chlorophenyl-2-methylpropionic acid, 4-chloro-alpha,alpha-dimethylphenylacetic acid, 2-p-chlorophenyl-2-methylpropionic acid, 4-chloro-.alpha.,.alpha.-dimethylphenylacetic acid, acmc-1bhp2, p-chlorophenylisobutyric acid, 2-methyl-2-4-chlorophenyl propanoic acid, 4-chloro-a,a-dimethylphenylacetic acid, 2-4-chloro phenyl-2-methylpropanoic acid
PubChem CID 80406
IUPAC Name 2-(4-chlorophenyl)-2-methylpropanoic acid
SMILES CC(C)(C1=CC=C(C=C1)Cl)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0527703
View Documents
Thermo Scientific Chemicals
L0527703
1 g
Each for $33.90
Only null left
 
AAL0527706
View Documents
Thermo Scientific Chemicals
L0527706
5 g
Each for $113.15
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

6258-30-6
198.646
SSFDAZXGUKDEAH-UHFFFAOYSA-N
80406
CC(C)(C1=CC=C(C=C1)Cl)C(=O)O
C10H11ClO2
MFCD00044814
2-4-chlorophenyl-2-methylpropanoic acid, 2-4-chlorophenyl-2-methylpropionic acid, 4-chloro-alpha,alpha-dimethylphenylacetic acid, 2-p-chlorophenyl-2-methylpropionic acid, 4-chloro-.alpha.,.alpha.-dimethylphenylacetic acid, acmc-1bhp2, p-chlorophenylisobutyric acid, 2-methyl-2-4-chlorophenyl propanoic acid, 4-chloro-a,a-dimethylphenylacetic acid, 2-4-chloro phenyl-2-methylpropanoic acid
2-(4-chlorophenyl)-2-methylpropanoic acid

Specifications

6258-30-6
C10H11ClO2
1 g
2-4-chlorophenyl-2-methylpropanoic acid, 2-4-chlorophenyl-2-methylpropionic acid, 4-chloro-alpha,alpha-dimethylphenylacetic acid, 2-p-chlorophenyl-2-methylpropionic acid, 4-chloro-.alpha.,.alpha.-dimethylphenylacetic acid, acmc-1bhp2, p-chlorophenylisobutyric acid, 2-methyl-2-4-chlorophenyl propanoic acid, 4-chloro-a,a-dimethylphenylacetic acid, 2-4-chloro phenyl-2-methylpropanoic acid
CC(C)(C1=CC=C(C=C1)Cl)C(=O)O
198.646
198.65
4-Chloro-alpha,alpha-dimethylphenylacetic acid
124°C to 129°C
MFCD00044814
2094056
SSFDAZXGUKDEAH-UHFFFAOYSA-N
2-(4-chlorophenyl)-2-methylpropanoic acid
80406
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 228-392-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only