missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-alpha,alpha-dimethylphenylacetic acid, 98%

Catalog No. AAL0527703
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0527703 1 g
AAL0527706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0527703 Supplier Thermo Scientific Chemicals Supplier No. L0527703
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 6258-30-6 | C10H11ClO2 | 198.646 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6258-30-6
Molecular Formula C10H11ClO2
Molecular Weight (g/mol) 198.646
MDL Number MFCD00044814
InChI Key SSFDAZXGUKDEAH-UHFFFAOYSA-N
Synonym 2-4-chlorophenyl-2-methylpropanoic acid, 2-4-chlorophenyl-2-methylpropionic acid, 4-chloro-alpha,alpha-dimethylphenylacetic acid, 2-p-chlorophenyl-2-methylpropionic acid, 4-chloro-.alpha.,.alpha.-dimethylphenylacetic acid, acmc-1bhp2, p-chlorophenylisobutyric acid, 2-methyl-2-4-chlorophenyl propanoic acid, 4-chloro-a,a-dimethylphenylacetic acid, 2-4-chloro phenyl-2-methylpropanoic acid
PubChem CID 80406
IUPAC Name 2-(4-chlorophenyl)-2-methylpropanoic acid
SMILES CC(C)(C1=CC=C(C=C1)Cl)C(=O)O

Specifications

Melting Point 124°C to 129°C
Quantity 1 g
Beilstein 2094056
Formula Weight 198.65
Percent Purity 98%
Chemical Name or Material 4-Chloro-alpha,alpha-dimethylphenylacetic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 228-392-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only