missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-7-(p-toluenesulfonyl)-7H-pyrrolo[2,3-d]pyrimidine 96.0+%, TCI America™

Catalog No. p-7125795
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C30361G 1 g
C30365G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C30361G Supplier TCI America Supplier No. C30361G

Chemical Identifiers

CAS 479633-63-1
Molecular Formula C13H10ClN3O2S
Molecular Weight (g/mol) 307.752
MDL Number MFCD09907939
InChI Key BTOJSYRZQZOMOK-UHFFFAOYSA-N
Synonym 4-Chloro-7-tosyl-7H-pyrrolo[2,3-d]pyrimidine
PubChem CID 44228967
IUPAC Name 4-chloro-7-(4-methylphenyl)sulfonylpyrrolo[2,3-d]pyrimidine
SMILES CC1=CC=C(C=C1)S(=O)(=O)N2C=CC3=C2N=CN=C3Cl

Specifications

Melting Point 147°C
Color White-Yellow
Quantity 1 g
Formula Weight 307.75
Percent Purity ≥96.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloro-7-(p-toluenesulfonyl)-7H-pyrrolo[2,3-d]pyrimidine