accessibility menu, dialog, popup

UserName

CAS RN 479633-63-1

IUPAC Name: 4-chloro-7-(4-methylbenzenesulfonyl)-7H-pyrrolo[2,3-d]pyrimidine
Synonyms: 4-chloro-7-(4-methylbenzenesulfonyl)pyrrolo[2,3-d]pyrimidine
Molecular Weight (g/mol): 307.75
Molecular Formula: C13H10ClN3O2S
InChi Key: BTOJSYRZQZOMOK-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)N1C=CC2=C(Cl)N=CN=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results