missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-nitrobenzophenone 98.0+%, TCI America™

Catalog No. C29065G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C29065G 5 g
C290625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C29065G Supplier TCI America Supplier No. C29065G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 56107-02-9
Molecular Formula C13H8ClNO3
Molecular Weight (g/mol) 261.66
MDL Number MFCD00007081
InChI Key YBDBYPQFIMSFJW-UHFFFAOYSA-N
Synonym 4-chloro-3-nitrobenzophenone, methanone, 4-chloro-3-nitrophenyl phenyl, benzophenone, 4-chloro-3-nitro, 4-chloro-3-nitrophenyl phenyl methanone, 3-nitro-4-chlorobenzophenone, chembl53270, 4-chloro-3-nitrophenyl-phenylmethanone, 4-chloro-3-nitro-phenyl-phenyl-methanone, 4-chloro-3-nitrophenyl phenyl ketone, acmc-20amlb
PubChem CID 41743
IUPAC Name (4-chloro-3-nitrophenyl)(phenyl)methanone
SMILES [O-][N+](=O)C1=CC(=CC=C1Cl)C(=O)C1=CC=CC=C1

Specifications

Melting Point 106°C
Color Yellow
Boiling Point 235°C
Quantity 5 g
Formula Weight 261.66
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloro-3-nitrobenzophenone
RTECSNumber DJ0425000
TSCA No