accessibility menu, dialog, popup

UserName

CAS RN 56107-02-9

IUPAC Name: (4-chloro-3-nitrophenyl)(phenyl)methanone
Synonyms: (4-chloro-3-nitrophenyl)(phenyl)methanone
Molecular Weight (g/mol): 261.66
Molecular Formula: C13H8ClNO3
InChi Key: YBDBYPQFIMSFJW-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(=CC=C1Cl)C(=O)C1=CC=CC=C1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)