missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-nitrobenzonitrile, 98%

CAS: 939-80-0 | C7H3ClN2O2 | 182.563 g/mol

$91.96 - $849.11

Chemical Identifiers

CAS 939-80-0
Molecular Formula C7H3ClN2O2
Molecular Weight (g/mol) 182.563
MDL Number MFCD00016987
InChI Key XBLPHYSLHRGMNW-UHFFFAOYSA-N
Synonym 3-nitro-4-chlorobenzonitrile, 4-chloro-3-nitro-benzonitrile, benzonitrile, 4-chloro-3-nitro, 4-chlor-3-nitrobenzonitril, 2-chloro-5-cyanonitrobenzene, 4-chlor-3-nitrobenzonitril czech, 4-chloro-3-nitro-benzonitril, chembl54162, 4-09-00-01227 beilstein handbook reference, 4-chloro-3-nitrobenzenecarbonitrile
PubChem CID 13655
IUPAC Name 4-chloro-3-nitrobenzonitrile
SMILES C1=CC(=C(C=C1C#N)[N+](=O)[O-])Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1264806
View Documents
Thermo Scientific Chemicals
A1264806
5 g
Each for $91.96
Only null left
 
AAA1264814
View Documents
Thermo Scientific Chemicals
A1264814
25 g
Each for $263.03
Only null left
 
AAA1264822
View Documents
Thermo Scientific Chemicals
A1264822
100 g
Each for $849.11
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

939-80-0
182.563
XBLPHYSLHRGMNW-UHFFFAOYSA-N
13655
C1=CC(=C(C=C1C#N)[N+](=O)[O-])Cl
C7H3ClN2O2
MFCD00016987
3-nitro-4-chlorobenzonitrile, 4-chloro-3-nitro-benzonitrile, benzonitrile, 4-chloro-3-nitro, 4-chlor-3-nitrobenzonitril, 2-chloro-5-cyanonitrobenzene, 4-chlor-3-nitrobenzonitril czech, 4-chloro-3-nitro-benzonitril, chembl54162, 4-09-00-01227 beilstein handbook reference, 4-chloro-3-nitrobenzenecarbonitrile
4-chloro-3-nitrobenzonitrile

Specifications

939-80-0
C7H3ClN2O2
5 g
3-nitro-4-chlorobenzonitrile, 4-chloro-3-nitro-benzonitrile, benzonitrile, 4-chloro-3-nitro, 4-chlor-3-nitrobenzonitril, 2-chloro-5-cyanonitrobenzene, 4-chlor-3-nitrobenzonitril czech, 4-chloro-3-nitro-benzonitril, chembl54162, 4-09-00-01227 beilstein handbook reference, 4-chloro-3-nitrobenzenecarbonitrile
C1=CC(=C(C=C1C#N)[N+](=O)[O-])Cl
182.563
182.57
4-Chloro-3-nitrobenzonitrile
98°C to 102°C
MFCD00016987
1639111
XBLPHYSLHRGMNW-UHFFFAOYSA-N
4-chloro-3-nitrobenzonitrile
13655
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 213-364-2

RTECSNumber : DI3050000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only