missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-nitrobenzonitrile, 98%

Catalog No. AAA1264806
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1264806 5 g
AAA1264814 25 g
AAA1264822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1264806 Supplier Thermo Scientific Chemicals Supplier No. A1264806

CAS: 939-80-0 | C7H3ClN2O2 | 182.563 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 939-80-0
Molecular Formula C7H3ClN2O2
Molecular Weight (g/mol) 182.563
MDL Number MFCD00016987
InChI Key XBLPHYSLHRGMNW-UHFFFAOYSA-N
Synonym 3-nitro-4-chlorobenzonitrile, 4-chloro-3-nitro-benzonitrile, benzonitrile, 4-chloro-3-nitro, 4-chlor-3-nitrobenzonitril, 2-chloro-5-cyanonitrobenzene, 4-chlor-3-nitrobenzonitril czech, 4-chloro-3-nitro-benzonitril, chembl54162, 4-09-00-01227 beilstein handbook reference, 4-chloro-3-nitrobenzenecarbonitrile
PubChem CID 13655
IUPAC Name 4-chloro-3-nitrobenzonitrile
SMILES C1=CC(=C(C=C1C#N)[N+](=O)[O-])Cl

Specifications

Melting Point 98°C to 102°C
Quantity 5 g
Beilstein 1639111
Formula Weight 182.57
Percent Purity 98%
Chemical Name or Material 4-Chloro-3-nitrobenzonitrile
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 213-364-2
RTECSNumber DI3050000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only