missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chloro-3-nitrobenzamide 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 154°C |
| Color | Yellow |
| Quantity | 5 g |
| Synonym | 3-nitro-4-chlorobenzamide, benzamide, 4-chloro-3-nitro, 4-chlor-3-nitrobenzamid czech, 4-chlor-3-nitrobenzamid, 3-nitro-4-chloro-benzamide, acmc-1c8t7, benzamide,4-chloro-3-nitro, 4-chloranyl-3-nitro-benzamide, 4-chloro-3-nitro benzamide |
| Formula Weight | 200.58 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4-Chloro-3-nitrobenzamide |
Safety and Handling
| EINECSNumber | (3)-1517 |
| RTECSNumber | CV2465000 |
| TSCA | Yes |