CAS RN 16588-06-0
IUPAC Name:
4-chloro-3-nitrobenzamide
Synonyms:
benzamide, 4-chloro-3-nitro-
Molecular Weight (g/mol):
200.58
Molecular Formula:
C7H5ClN2O3
InChi Key:
CGXRJCDXGJRBHV-UHFFFAOYSA-N
SMILES:
NC(=O)C1=CC=C(Cl)C(=C1)[N+]([O-])=O
1
–
1
of
1
results
1
4-Chloro-3-nitrobenzamide 98.0+%, TCI America™
CAS: 16588-06-0 Molecular Formula: C7H5ClN2O3 Molecular Weight (g/mol): 200.578 MDL Number: MFCD00017134 InChI Key: CGXRJCDXGJRBHV-UHFFFAOYSA-N PubChem CID: 27942 IUPAC Name: 4-chloro-3-nitrobenzamide SMILES: C1=CC(=C(C=C1C(=O)N)[N+](=O)[O-])Cl