missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-nitrobenzamide 98.0+%, TCI America™

Catalog No. C31761G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C31761G 1 g
C31765G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C31761G Supplier TCI America Supplier No. C31761G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 16588-06-0
Molecular Formula C7H5ClN2O3
Molecular Weight (g/mol) 200.578
MDL Number MFCD00017134
InChI Key CGXRJCDXGJRBHV-UHFFFAOYSA-N
PubChem CID 27942
IUPAC Name 4-chloro-3-nitrobenzamide
SMILES C1=CC(=C(C=C1C(=O)N)[N+](=O)[O-])Cl

Specifications

Melting Point 154°C
Color Yellow
Quantity 1 g
Synonym 3-nitro-4-chlorobenzamide, benzamide, 4-chloro-3-nitro, 4-chlor-3-nitrobenzamid czech, 4-chlor-3-nitrobenzamid, 3-nitro-4-chloro-benzamide, acmc-1c8t7, benzamide,4-chloro-3-nitro, 4-chloranyl-3-nitro-benzamide, 4-chloro-3-nitro benzamide
Formula Weight 200.58
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloro-3-nitrobenzamide
EINECSNumber (3)-1517
RTECSNumber CV2465000
TSCA Yes