missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-nitrobenzaldehyde, 97%

Catalog No. p-7021276
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1354909 10 g
AAA1354918 50 g
AAA1354930 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1354909 Supplier Thermo Scientific Chemicals Supplier No. A1354909
Only null left

CAS: 16588-34-4 | C7H4ClNO3 | 185.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 16588-34-4
Molecular Formula C7H4ClNO3
Molecular Weight (g/mol) 185.56
MDL Number MFCD00007078
InChI Key HETBKLHJEWXWBM-UHFFFAOYSA-N
Synonym 3-nitro-4-chlorobenzaldehyde, benzaldehyde, 4-chloro-3-nitro, chembl51664, 4-chloro-3-nitro-benzaldehyde, pubchem14274, acmc-1c7bn, 3nitro-4-chlorobenzaldehyde, asischem r36965, dsstox_cid_24846, dsstox_rid_80526
PubChem CID 85505
IUPAC Name 4-chloro-3-nitrobenzaldehyde
SMILES [O-][N+](=O)C1=CC(C=O)=CC=C1Cl

Specifications

Melting Point 61°C to 65°C
Quantity 10 g
Beilstein 778323
Sensitivity Air Sensitive
Formula Weight 185.57
Percent Purity 97%
Chemical Name or Material 4-Chloro-3-nitrobenzaldehyde
Hazard Category H315-H317-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P272-P280-P302+P352-P304+P340-P305+P351+P338-P312-P333+P313-P362-P501c
EINECSNumber 240-645-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only