accessibility menu, dialog, popup

UserName

CAS RN 16588-34-4

IUPAC Name: 4-chloro-3-nitrobenzaldehyde
Synonyms: 4-chloro-3-nitrobenzaldehyde
Molecular Weight (g/mol): 185.56
Molecular Formula: C7H4ClNO3
InChi Key: HETBKLHJEWXWBM-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(C=O)=CC=C1Cl
Percent Purity 
  • (2)
  • (6)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (2)
  • (6)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)