missing translation for 'onlineSavingsMsg'
Learn More

4'-Chloro-3'-nitroacetophenone 98.0+%, TCI America™

Supplier:  TCI America C232125G

 View more versions of this product

Catalog No. C232125G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

4′-Chloro-3′-nitroacetophenone
99°C
MFCD00007083
4'-chloro-3'-nitroacetophenone, 1-4-chloro-3-nitrophenyl ethanone, 4-chloro-3-nitroacetophenone, ethanone, 1-4-chloro-3-nitrophenyl, 1-4-chloro-3-nitrophenyl ethan-1-one, unii-gkv85f2zxl, acetophenone, 4'-chloro-3'-nitro, gkv85f2zxl, 4-chloro-3'-nitroacetophenone, 3'-nitro-4'-chloro acetophenone
CC(=O)C1=CC=C(Cl)C(=C1)[N+]([O-])=O
199.59
199.59
Crystalline Powder
5465-65-6
C8H6ClNO3
25 g
YEVPHFIFGUWSMG-UHFFFAOYSA-N
1-(4-chloro-3-nitrophenyl)ethan-1-one
79596
≥98.0% (GC)

Chemical Identifiers

5465-65-6
199.59
YEVPHFIFGUWSMG-UHFFFAOYSA-N
79596
CC(=O)C1=CC=C(Cl)C(=C1)[N+]([O-])=O
C8H6ClNO3
MFCD00007083
4'-chloro-3'-nitroacetophenone, 1-4-chloro-3-nitrophenyl ethanone, 4-chloro-3-nitroacetophenone, ethanone, 1-4-chloro-3-nitrophenyl, 1-4-chloro-3-nitrophenyl ethan-1-one, unii-gkv85f2zxl, acetophenone, 4'-chloro-3'-nitro, gkv85f2zxl, 4-chloro-3'-nitroacetophenone, 3'-nitro-4'-chloro acetophenone
1-(4-chloro-3-nitrophenyl)ethan-1-one
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions