missing translation for 'onlineSavingsMsg'
Learn More

4'-Chloro-3'-nitroacetophenone 98.0+%, TCI America™

Catalog No. p-7125407
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C23215G 5 g
C232125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C23215G Supplier TCI America Supplier No. C23215G

Chemical Identifiers

CAS 5465-65-6
Molecular Formula C8H6ClNO3
Molecular Weight (g/mol) 199.59
MDL Number MFCD00007083
InChI Key YEVPHFIFGUWSMG-UHFFFAOYSA-N
Synonym 4'-chloro-3'-nitroacetophenone, 1-4-chloro-3-nitrophenyl ethanone, 4-chloro-3-nitroacetophenone, ethanone, 1-4-chloro-3-nitrophenyl, 1-4-chloro-3-nitrophenyl ethan-1-one, unii-gkv85f2zxl, acetophenone, 4'-chloro-3'-nitro, gkv85f2zxl, 4-chloro-3'-nitroacetophenone, 3'-nitro-4'-chloro acetophenone
PubChem CID 79596
IUPAC Name 1-(4-chloro-3-nitrophenyl)ethan-1-one
SMILES CC(=O)C1=CC=C(Cl)C(=C1)[N+]([O-])=O

Specifications

Melting Point 99°C
Quantity 5 g
Formula Weight 199.59
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Chloro-3′-nitroacetophenone