missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chloro-3-methoxybenzoic Acid 98.0+%, TCI America™
$72.70 - $208.78
Chemical Identifiers
| CAS | 85740-98-3 |
|---|---|
| Molecular Formula | C8H7ClO3 |
| Molecular Weight (g/mol) | 186.591 |
| MDL Number | MFCD00269640 |
| InChI Key | OXUUNDMDOOXPKY-UHFFFAOYSA-N |
| Synonym | 4-Chloro-m-anisic Acid |
| PubChem CID | 2063378 |
| IUPAC Name | 4-chloro-3-methoxybenzoic acid |
| SMILES | COC1=C(C=CC(=C1)C(=O)O)Cl |
Chemical Identifiers
| 85740-98-3 | |
| 186.591 | |
| OXUUNDMDOOXPKY-UHFFFAOYSA-N | |
| 2063378 | |
| COC1=C(C=CC(=C1)C(=O)O)Cl |
| C8H7ClO3 | |
| MFCD00269640 | |
| 4-Chloro-m-anisic Acid | |
| 4-chloro-3-methoxybenzoic acid |
Specifications
| 85740-98-3 | |
| White | |
| MFCD00269640 | |
| 4-Chloro-m-anisic Acid | |
| COC1=C(C=CC(=C1)C(=O)O)Cl | |
| 186.591 | |
| 186.59 | |
| Crystalline Powder |
| 219°C | |
| C8H7ClO3 | |
| 1 g | |
| OXUUNDMDOOXPKY-UHFFFAOYSA-N | |
| 4-chloro-3-methoxybenzoic acid | |
| 2063378 | |
| ≥98.0% (GC,T) | |
| 4-Chloro-3-methoxybenzoic Acid |
Safety and Handling
TSCA : No