accessibility menu, dialog, popup

UserName

CAS RN 85740-98-3

H3COClOO

CAS RN 85740-98-3

IUPAC Name: 4-chloro-3-methoxybenzoate
Synonyms: 4-chloro-3-methoxybenzoate
Molecular Weight (g/mol): 185.58
Molecular Formula: C8H6ClO3
InChi Key: OXUUNDMDOOXPKY-UHFFFAOYSA-M
SMILES: COC1=CC(=CC=C1Cl)C([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results