missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-methoxybenzoic Acid 98.0+%, TCI America™

Catalog No. C33045G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C33045G 5 g
C33041G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C33045G Supplier TCI America Supplier No. C33045G
Only null left

Chemical Identifiers

CAS 85740-98-3
Molecular Formula C8H7ClO3
Molecular Weight (g/mol) 186.591
MDL Number MFCD00269640
InChI Key OXUUNDMDOOXPKY-UHFFFAOYSA-N
Synonym 4-Chloro-m-anisic Acid
PubChem CID 2063378
IUPAC Name 4-chloro-3-methoxybenzoic acid
SMILES COC1=C(C=CC(=C1)C(=O)O)Cl

Specifications

Melting Point 219°C
Color White
Quantity 5 g
Formula Weight 186.59
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloro-3-methoxybenzoic Acid